Order Entry
Poland
ContactUsLinkComponent
Anti-COL4A1 Mouse Monoclonal Antibody clone: [2F11] (PE (Phycoerythrin))
Anti-COL4A1 Mouse Monoclonal Antibody clone: [2F11] (PE (Phycoerythrin))
Catalog # PRSI98-530
Supplier:  ProSci Inc.
Anti-COL4A1 Mouse Monoclonal Antibody clone: [2F11] (PE (Phycoerythrin))
Catalog # PRSI98-530
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    collagen type IV alpha 1 chain
  • Antigen symbol:
    COL4A1
  • Clonality:
    Monoclonal
  • Clone:
    [2F11]
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    1282;1284;1285;1286;1287;1288
  • Antigen synonyms:
    COLIV
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS/NaN₃ and a stabilising agent
  • Storage temperature:
    Store vial at 2...8 °C
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    20
  • Immunogen:
    Native human type IV collagen
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: COL4A1
Clonality: Monoclonal
Clone: [2F11]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human