Anti-CYP2D6 Rabbit Polyclonal Antibody
PRSI14-731
New Product
- Antibody type:Primary
- Antigen name:cytochrome P450, family 2, subfamily D, polypeptide 6
- Antigen symbol:CYP2D6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1565
- Antigen synonyms:CYP2D@|P450C2D|CYP2D7BP|P450-DB1|CYP2DL1|P450DB1|MGC120390|RP4-669P10.2|CYP2D7AP|CYP2D8P2|CYP2D7P2|CYP2D6|CYPIID6|MGC120389|CYP2D|CPD6
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human CYP2D6 (NP_000097.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat