Anti-ALPL Rabbit Polyclonal Antibody
Catalog # PRSI14-397
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:alkaline phosphatase, liver/bone/kidney
- Antigen symbol:ALPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:249
- Antigen synonyms:Tissue-nonspecific ALP|ALPL|Liver alkaline phosphatase|Alkaline phosphatase liver|Glycerophosphatase|TNSALP|AP-TNAP|Alkaline phosphomonoesterase|TNAP|HOPS|APTNAP
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 150-250 of human ALPL (NP_000469.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ALPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat