Anti-CGA Rabbit Polyclonal Antibody
Catalog # PRSI14-394
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:glycoprotein hormones, alpha polypeptide
- Antigen symbol:CGA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1081
- Antigen synonyms:Follitropin alpha chain|Glycoprotein hormones alpha chain|Choriogonadotropin alpha chain|Luteinizing hormone alpha chain|CG-alpha|Chorionic gonadotrophin subunit alpha|Thyrotropin alpha chain|FSH-alpha|Follicle-stimulating hormone alpha chain|LSH-alpha|TSH-alpha|Lutropin alpha chain|CGA|Anterior pituitary glycoprotein hormones common subunit alpha|Thyroid-stimulating hormone alpha chain
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 25-116 of human CGA (NP_000726.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse