Anti-SLC16A7 Rabbit Polyclonal Antibody
Catalog # PRSI14-392
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 16 (monocarboxylate transporter), member 7
- Antigen symbol:SLC16A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9194
- Antigen synonyms:solute carrier family 16 (monocarboxylic acid transporters)|member 7|monocarboxylate transporter 2|SLC16A7|MCT2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human SLC16A7 (NP_001257551.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC16A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat