Anti-SLC25A19 Rabbit Polyclonal Antibody
PRSI14-382
New Product
- Antibody type:Primary
- Antigen name:Mitochondrial thiamine pyrophosphate carrier
- Antigen symbol:SLC25A19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:60386
- Antigen synonyms:SLC25A19|Mitochondrial uncoupling protein 1|MUP1|Solute carrier family 25 member 19|Mitochondrial thiamine pyrophosphate carrier|DNC
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human SLC25A19 (NP_068380.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC25A19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat