Anti-CHRM1 Rabbit Polyclonal Antibody
Catalog # PRSI14-364
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:cholinergic receptor, muscarinic 1
- Antigen symbol:CHRM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1128
- Antigen synonyms:HM1|MGC30125|M1R|CHRM1|cholinergic receptor|M1|muscarinic acetylcholine receptor M1|muscarinic 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human CHRM1 (NP_000729.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CHRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat