Anti-C8orf4 Rabbit Polyclonal Antibody
Catalog # PRSI22-076
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Transcriptional and immune response regulator
- Antigen symbol:C8orf4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:56892
- Antigen synonyms:TC-1|transcriptional and immune response regulator|thyroid cancer protein 1|TC1|human thyroid cancer 1|C8orf4|protein C8orf4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-106 of human C8orf4 (NP_064515.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: C8orf4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat