Anti-SLC32A1 Rabbit Polyclonal Antibody
Catalog # PRSI18-955
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 32 member 1
- Antigen symbol:SLC32A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:140679
- Antigen synonyms:vesicular inhibitory amino acid transporter|GABA and glycine transporter|VGAT|solute carrier family 32 (GABA vesicular transporter)|hVIAAT|member 1|VIAAT|vesicular GABA transporter
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human SLC32A1.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat