Anti-ITGB3 Mouse Monoclonal Antibody [clone: VI-PL2]
PRSI76-175
- Antibody type:Primary
- Antigen name:integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
- Antigen symbol:ITGB3
- Clonality:Monoclonal
- Clone:VI-PL2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:P05106
- Antigen synonyms:ITGB3
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional
- Purification:The monoclonal antibody was purified utilizing affinity chromatography.
- Pk:0,1 mg
The VI-PL2 monoclonal antibody reacts with CD61, a type I integral transmembrane glycoprotein also known as integrin beta 3. CD61 forms a complex with CD41 (platelet gpIIb) and CD51 (integrin alphaV). It is expressed on osteoclasts, fibroblasts, platelets, macrophages, and megakaryocytes and appears to bind to fibronectin, vitronectin, thrombospondin, and fibrinogen and vWF. The VI-PL2 antibody is reported to be cross-reactive with baboon, rhesus, and cynomolgus.
Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human