Anti-ITGB3 Mouse Monoclonal Antibody [clone: VI-PL2] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-177
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
- Antigen symbol:ITGB3
- Clonality:Monoclonal
- Clone:VI-PL2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P05106
- Antigen synonyms:ITGB3
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The VI-PL2 monoclonal antibody reacts with CD61, a type I integral transmembrane glycoprotein also known as integrin beta 3. CD61 forms a complex with CD41 (platelet gpIIb) and CD51 (integrin alphaV). It is expressed on osteoclasts, fibroblasts, platelets, macrophages, and megakaryocytes and appears to bind to fibronectin, vitronectin, thrombospondin, and fibrinogen and vWF. The VI-PL2 antibody is reported to be cross-reactive with baboon, rhesus, and cynomolgus.
Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human