Anti-RPL12 Rabbit Polyclonal Antibody
Catalog # PRSI58-375
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:60S ribosomal protein L12
- Antigen symbol:RPL12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:71 - 100 aa
- Form:Liquid
- Gene ID:6136
- Modification:None
- Molecular weight:18 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RPL12 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 71-100 amino acids from the Central region of human RPL12
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: RPL12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 71-100
Host: Rabbit
Isotype: Ig
Reactivity: Human