Anti-GTF3C2 Rabbit Polyclonal Antibody
Catalog # PRSI58-356
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 3C polypeptide 2
- Antigen symbol:GTF3C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:141 - 170 aa
- Form:Liquid
- Gene ID:2976
- Antigen synonyms:transcription factor IIIC 110 kDa subunit|TFIIIC-BETA|general transcription factor 3C polypeptide 2|TFIIIC 110 kDa subunit|transcription factor IIIC subunit beta|TFIIIC110|TF3C-beta
- Modification:None
- Molecular weight:101 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GTF3C2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 141-170 amino acids from the N-terminal region of human GTF3C2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GTF3C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 141-170
Host: Rabbit
Isotype: Ig
Reactivity: Human