Anti-STAT5B Rabbit Polyclonal Antibody
Catalog # PRSI58-172
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: STAT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 760-787
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Antibody type:Primary
- Epitope:760 - 787 aa
- Form:Liquid
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Host:Rabbit
- Concentration:2 mg/ml
- Molecular weight:90 kDa
- Shipping temperature:4 °C
- Antigen name:Signal transducer and activator of transcription 5B
- Cat. No.:PRSI58-172
- Gene ID:6777
- Immunogen:This STAT5b antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 760-787 amino acids from the C-terminal region of human STAT5b
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:STAT5B



