Order Entry
Poland
ContactUsLinkComponent
 
Anti-ADAMTS2 Rabbit Polyclonal Antibody
Catalog # PRSI13-548
Supplier:  ProSci Inc.
Anti-ADAMTS2 Rabbit Polyclonal Antibody
Catalog # PRSI13-548
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 2
  • Antigen symbol:
    ADAMTS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    216725
  • Antigen synonyms:
    ADAM-TS 2|ADAMTS-2|pNPI|procollagen N-endopeptidase|A disintegrin and metalloproteinase with thrombospondin motifs 2|PCINP|procollagen I/II amino propeptide-processing enzyme|a disintegrin and metalloproteinase with thrombospondin repeats|2|ADAM-TS2|procollagen N-proteinase|a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif|PC I-NP
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant protein of mouse ADAMTS2
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: ADAMTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat