Anti-MAPK8 Rabbit Polyclonal Antibody
Catalog # PRSI13-131
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 8
- Antigen symbol:MAPK8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5599
- Antigen synonyms:SAPK1|Stress-activated protein kinase JNK1|MAPK 8|PRKM8|Stress-activated protein kinase 1c|JNK-46|MAP kinase 8|Mitogen-activated protein kinase 8|SAPK1C|JNK1|SAPK1c|c-Jun N-terminal kinase 1|MAPK8
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 245-345 of human JNK1 (NP_620635.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MAPK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse