Order Entry
Poland
ContactUsLinkComponent
Anti-CCR5 Recombinant Antibody [Clone: E11/19]
Anti-CCR5 Recombinant Antibody [Clone: E11/19]
Catalog # PRSI24-784
Supplier:  ProSci Inc.
Anti-CCR5 Recombinant Antibody [Clone: E11/19]
Catalog # PRSI24-784
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-C motif chemokine receptor 5
  • Antigen symbol:
    CCR5
  • Clonality:
    Recombinant
  • Clone:
    E11/19
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    1234
  • Antigen synonyms:
    CD195|C-C CKR-5|C-C chemokine receptor type 5|CC-CKR-5|Ser349|pS349|Ser-349|CHEMR13|CCR-5|p349|HIV-1 fusion coreceptor
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizing mice with a synthetic phosphopeptide (CEAPERA(pS)(pS)VYTR(pS)TGEQI(pS)VGL) corresponding to the C-terminal region of human CCR5
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: C-C motif chemokine receptor 5
Clonality: Recombinant
Clone: E11/19
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human