Anti-Tnfsf9 Recombinant Antibody [Clone: AT113-2]
PRSI24-612
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 9
- Antigen symbol:Tnfsf9
- Clonality:Recombinant
- Clone:AT113-2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:21950
- Antigen synonyms:CDw137L|receptor 4-1BB ligand|homolog of mouse 4-1BB-L|41BBL|CD137L|tumor necrosis factor (ligand) superfamily|Tumor necrosis factor ligand superfamily member 9.|4-1BB ligand|TNFSF9|tumor necrosis factor (ligand) superfamily member 9|tumor necrosis factor ligand 5A|member 9
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising LOU rats with soluble recombinant 4-1BBL:Fc fusion protein
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: tumor necrosis factor (ligand) superfamily, member 9
Clonality: Recombinant
Clone: AT113-2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Mouse