Order Entry
Poland
ContactUsLinkComponent
Anti-MSLN Recombinant Antibody [Clone: IC14-30]
Anti-MSLN Recombinant Antibody [Clone: IC14-30]
Catalog # PRSI31-541
Supplier:  ProSci Inc.
Anti-MSLN Recombinant Antibody [Clone: IC14-30]
Catalog # PRSI31-541
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    mesothelin
  • Antigen symbol:
    MSLN
  • Clonality:
    Recombinant
  • Clone:
    IC14-30
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    10232
  • Antigen synonyms:
    Pre-pro-megakaryocyte-potentiating factor|CAK1 antigen|MSLN
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    The original antibody was generated by intraperitoneally immunizing BALB/c mice with the purified recombinant mesothelin protein
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: mesothelin
Clonality: Recombinant
Clone: IC14-30
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human