Anti-C3 Recombinant Antibody [Clone: h-R3 (Nimotuzumab)]
PRSI24-489
- Antibody type:Primary
- Antigen name:complement C3
- Antigen symbol:C3
- Clonality:Recombinant
- Clone:h-R3 (Nimotuzumab)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:718
- Antigen synonyms:ERBB-D3|PIG61|HER1|mENA|epidermal growth factor|proto-oncogene c-ErbB-1|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody we generated by humanisation of the murine antibody R3, raised against EGFR purified from placenta membranes and A431 tumor
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Type: Primary,Biosimilars
Antigen: complement C3
Clonality: Recombinant
Clone: h-R3 (Nimotuzumab)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: human