Order Entry
Poland
ContactUsLinkComponent
Orthoboric acid 40 g/l in aqueous solution
Orthoboric acid 40 g/l in aqueous solution
  5792.5000
 :  
undefined
Orthoboric acid 40 g/l in aqueous solution
  5792.5000
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

 

 

  • Assay:
    3.9 - 4.1 %

 

 

 :
 :1.0174 g/cm³
 :61.83 g/mol
 :Ambient
 :233-139-2
 :10043-35-3
 :B(OH)₃
 :MFCD00011337