Order Entry
Poland
ContactUsLinkComponent
Potassium dihydrogen phosphate 1/15 mol/l in aqueous solution, Supelco®
Potassium dihydrogen phosphate 1/15 mol/l in aqueous solution, Supelco®
Catalog # 1.04875.1000
Supplier:  Merck
CAS Number:  
undefined
Potassium dihydrogen phosphate 1/15 mol/l in aqueous solution, Supelco®
Catalog # 1.04875.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Density:
    1.001 g/cm3

Specification Test Results

About this item

 :
 :136.09 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-913-4
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401