Hexafluorosilicic acid 23% (w/w) in aqueous solution
Catalog # 69109.36
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Fluosilicic acid
CAS Number:16961-83-4
:
Formula:H₂F₆Si
MDL Number:MFCD00036289
:144.09 g/mol
Density:1.223
EINECS:241-034-8
UN:1778
:GHS05
:GHS07
Storage Temperature:Ambient
ADR:8 II
Merck Index:14,4182