1,7-Dimethylxanthine
Catalog # PROOMM0494.04
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:180.17 g/mol
EINECS:210-271-9
:GHS07
CAS Number:611-59-6
Formula:C₇H₈N₄O₂
MDL Number:MFCD00005727