
Congo red, Supelco®
Catalog # 1.01340.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:25 g/l (20 °C)
- Melting Point:360 °C
- Molar Mass:696.68 g/mol
- Bulk Density:600 - 700 kg/m3
Specification Test Results
About this item
:
:696.67 g/mol
Merck Index:12,02562
Melting Pt:> 360 °C
Storage Temperature:Ambient
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:209-358-4
:GHS08
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028