
(±)Epibatidine ≥99% (by HPLC)
Catalog # ENZOBMLC1140005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥99% (HPLC)
Specification Test Results
About this item
:
:208.7 g/mol
UN:2811
ADR:6.1,I
:GHS06
CAS Number:140111-52-0
Formula:C₁₁H₁₃ClN₂