Order Entry
Poland
ContactUsLinkComponent
Clindamycin hydrochloride ≥95% (by HPLC)
Clindamycin hydrochloride ≥95% (by HPLC)
Catalog # ENZOBMLA1900100
Supplier:  Enzo Life Sciences
CAS Number:  
Clindamycin hydrochloride ≥95% (by HPLC)
Catalog # ENZOBMLA1900100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :461.45 g/mol
Storage Temperature:Refrigerator
CAS Number:21462-39-5
Formula:C₁₈H₃₃ClN₂O₅S·HCl
MDL Number:MFCD07793327