ODN 1668 (synthetic), TLRgrade™, sterile, lyophilised
Catalog # ENZOALX746051M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Source:Synthetic
- Formulation:Lyophilised. Sterile.
- Purity:Activity and endotoxin tested - TLRgrade™
- Endotoxin content:<0.002 EU/µg
- Quantity:157.1 nmol
Specification Test Results
About this item
:
:6.383 g/mol
Storage Temperature:Refrigerator