Polyinosinic-polycytidylic acid potassium salt (synthetic), TLRgrade™
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:31852-29-6
:
Formula:(C₁₀H₁₀N₄NaO₇P)x·(C₉H₁₁N₃NaO₇P)x
Storage Temperature:Refrigerator
Specification Test Results
- Source:Synthetic
- Purity :Activity and endotoxin tested - TLRgrade™
- Endotoxin Content:<0.002 EU/µg
- Formulation:Lyophilised, sterile
Specification Test Results
Frequently Bought Together