Order Entry
Poland
ContactUsLinkComponent
 
ODN 1826 (synthetic), TLRgrade™
Catalog # ENZOALX746002C100
Supplier:  Enzo Life Sciences
CAS Number:  
ODN 1826 (synthetic), TLRgrade™
Catalog # ENZOALX746002C100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Formulation:
    Lyophilised. Sterile.
  • Purity:
    Activity and endotoxin tested - TLRgrade™
  • Endotoxin content:
    <0.002 EU/µg
  • Quantity:
    15 nmol (~96µg)

Specification Test Results

About this item

 :
 :6.383 g/mol
Storage Temperature:Refrigerator