
DAF-2 ≥98% (by HPLC)
Catalog # ENZOALX620052M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Concentration:~5 mM
- Formulation:Dissolved in 0.55 ml DMSO
- Purity:≥98% (HPLC)
Specification Test Results
About this item
:
:364.34 g/mol
EINECS:200-664-3
:GHS07
Storage Temperature:Freezer
CAS Number:205391-01-1
Formula:C₂₀H₁₆N₂O₅
MDL Number:MFCD01866182