Order Entry
Poland
ContactUsLinkComponent
(2S,4R)-4-Methylglutamic acid ≥97%
(2S,4R)-4-Methylglutamic acid ≥97%
Catalog # ENZOALX550186M005
Supplier:  Enzo Life Sciences
CAS Number:  
(2S,4R)-4-Methylglutamic acid ≥97%
Catalog # ENZOALX550186M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :161.2 g/mol
Storage Temperature:Freezer
CAS Number:31137-74-3
Formula:C₆H₁₁NO₄
MDL Number:MFCD00937765