Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Noc-7
Noc-7 ≥96% (by HPLC)
Noc-7 ≥96% (by HPLC)
:
Enzo Life Sciences
Noc-7 ≥96% (by HPLC)
:
Enzo Life Sciences
:
146724-84-7
:
:
C₅H₁₄N₄O₂
:
162.2 g/mol
:
GHS07
:
Freezer
Purity:
≥96% (HPLC)
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Serological pipettes, Stripette™
: Corning
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF