GEA 5024 ≥97% (by HPLC)
Catalog # ENZOALX430005M010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥97% (HPLC)
Specification Test Results
About this item
:
:247.1 g/mol
Storage Temperature:Refrigerator
CAS Number:144575-27-9
Formula:C₈H₇ClN₄O·HCl