Prodigiosin ≥95% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:82-89-3
:
Frequently Bought Together
Cell culture inserts, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
VWR® Cell Culture Dishes
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
J.T.Baker® Plate, SRW Magnetic
Supplier: Avantor
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Cell culture plates and Multidishes, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
Sampling spoons, long handle, disposable, LaboPlast®/SteriPlast®
Supplier: BURKLE