Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enniatin B
Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
ENZOALX380007M001
:
Enzo Life Sciences
:
Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
ENZOALX380007M001
:
Enzo Life Sciences
:
Pk:
1 mg
Seq:
2070
:
:
639.8 g/mol
:
Freezer
:
917-13-5
:
C₃₃H₅₇N₃O₉