Order Entry
Poland
ContactUsLinkComponent
Xanthorrhizol ≥97% (by HPLC)
Health
Xanthorrhizol ≥97% (by HPLC)
Catalog # ENZOALX350263M001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Health
Xanthorrhizol ≥97% (by HPLC)
Catalog # ENZOALX350263M001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity:
    ≥97% (HPLC)
  • Identity:
    Determined by ¹H-NMR, ¹³C-NMR and MS
  • Source:
    Synthetic

Specification Test Results

About this item

 :
 :218.3 g/mol
 :GHS08
Storage Temperature:Freezer
CAS Number:30199-26-9
Formula:C₁₅H₂₂O