Order Entry
Poland
ContactUsLinkComponent
 
D-myo-Inositol-1-monophosphate dipotassium salt ≥98% (by HPLC)
  ENZOALX307026M001
 :  
undefined
D-myo-Inositol-1-monophosphate dipotassium salt ≥98% (by HPLC)
  ENZOALX307026M001
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

  • Purity:
    ≥98% by HPLC
  • Identity:
    Determined by ¹H-NMR, ³¹P-NMR and MS

 

 

 :
 :336.3 g/mol
 :Freezer
 :573-35-3
 :C₆H₁₁O₉P·2K