Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
UPF1035
UPF1035 ≥95% (by HPLC)
UPF1035 ≥95% (by HPLC)
ENZOALX270498M005
:
ENZO LIFE SCIENCES
:
UPF1035 ≥95% (by HPLC)
ENZOALX270498M005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
:
:
265.3 g/mol
:
Freezer
:
370872-09-6
:
C₁₆H₁₁NO₃