N-ω-Hydroxy-L-norarginine acetate salt ≥97% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:189302-40-7
:
Formula:C₅H₁₂N₄O₃.CH₃COOH
MDL Number:MFCD01318777
:236.3 g/mol
Storage Temperature:Zamra·arka
Frequently Bought Together