Order Entry
Poland
ContactUsLinkComponent
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline ≥97% (by 1H-NMR)
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline ≥97% (by 1H-NMR)
Catalog # ENZOALX270448M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline ≥97% (by 1H-NMR)
Catalog # ENZOALX270448M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :272.31 g/mol
Storage Temperature:Freezer
CAS Number:396129-53-6
Formula:C₁₇H₁₂N₄
MDL Number:MFCD00800758