CDK1/2 Inhibitor III ≥95% (by HPLC)
Catalog # ENZOALX270442M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:425.4 g/mol
Storage Temperature:Freezer
CAS Number:443798-55-8
Formula:C₁₅H₁₃F₂N₇O₂S₂