
Pioglitazone ≥97% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:111025-46-8
:
Formula:C₁₉H₂₀N₂O₃S
MDL Number:MFCD00865504
:356.44 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity:≥97% (HPLC)
- Identity:Confirmed by ¹H-NMR
Specification Test Results
Frequently Bought Together