Order Entry
Poland
ContactUsLinkComponent
TIQ-A ≥96% (by HPLC)
TIQ-A ≥96% (by HPLC)
Catalog # ENZOALX270365M005
Supplier:  Enzo Life Sciences
CAS Number:  
TIQ-A ≥96% (by HPLC)
Catalog # ENZOALX270365M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :201.2 g/mol
Storage Temperature:Refrigerator
CAS Number:420849-22-5
Formula:C₁₁H₇NOS