Order Entry
Poland
ContactUsLinkComponent
Chlorpromazine hydrochloride ≥98% (by HPLC)
Chlorpromazine hydrochloride ≥98% (by HPLC)
Catalog # ENZOALX270171G005
Supplier:  Enzo Life Sciences
CAS Number:  
Chlorpromazine hydrochloride ≥98% (by HPLC)
Catalog # ENZOALX270171G005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :355.32 g/mol
CAS Number:69-09-0
Formula:C₁₇H₁₉ClN₂S
MDL Number:MFCD00012654