Order Entry
Poland
ContactUsLinkComponent
Tyrphostin AG 527 ≥98% (by HPLC)
Tyrphostin AG 527 ≥98% (by HPLC)
Catalog # ENZOALX270141M025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Tyrphostin AG 527 ≥98% (by HPLC)
Catalog # ENZOALX270141M025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :308.3 g/mol
Storage Temperature:Freezer
CAS Number:133550-32-0
Formula:C₁₈H₁₆N₂O₃