Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Milrinone
Milrinone ≥97% (by TLC)
Milrinone ≥97% (by TLC)
Milrinone ≥97% (by TLC)
Catalog # ENZOALX270083M005
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Milrinone ≥97% (by TLC)
Catalog # ENZOALX270083M005
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
211.22 g/mol
CAS Number:
78415-72-2
Formula:
C₁₂H₉N₃O
MDL Number:
MFCD00133539
Product Details