Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ac-WEHD-pNA
Ac-WEHD-pNA ≥98% (by HPLC)
Ac-WEHD-pNA ≥98% (by HPLC)
Ac-WEHD-pNA ≥98% (by HPLC)
Catalog # ENZOALX260082M005
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Ac-WEHD-pNA ≥98% (by HPLC)
Catalog # ENZOALX260082M005
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
747.70 g/mol
Storage Temperature:
Freezer
Formula:
C₃₄H₃₇N₉O₁₁
Product Details