Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
R-(-)-Rolipram
R-(-)-Rolipram ≥98% (by HPLC)
R-(-)-Rolipram ≥98% (by HPLC)
R-(-)-Rolipram ≥98% (by HPLC)
ENZOBMLPD1770005
:
Enzo Life Sciences
:
R-(-)-Rolipram ≥98% (by HPLC)
ENZOBMLPD1770005
:
Enzo Life Sciences
:
Pk:
5 mg
Seq:
2210
:
:
275.34 g/mol
:
Ambient
:
85416-75-7
:
MFCD03093861
:
C₁₆H₂₁NO₃
:
MFCD03093860