Order Entry
Poland
ContactUsLinkComponent
Ansamitocin P-3 ≥93% (by HPLC)
Ansamitocin P-3 ≥93% (by HPLC)
Catalog # ENZOBMLT1210001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Ansamitocin P-3 ≥93% (by HPLC)
Catalog # ENZOBMLT1210001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :635.14 g/mol
Storage Temperature:Freezer
CAS Number:66547-09-9
Formula:C₃₂H₄₃ClN₂O₉
MDL Number:MFCD00274586